Learn More
PurSwab™ Small Microfiber Swab
Catalog No. 22146662
Change view
Click to view available options
Length (English):
3 in.
Catalog No. Length (English)
22-146-662 3 in.
1 options
Catalog No. 22-146-662 Supplier PurSwab™ Supplier No. 3130
Add to Cart
Add to Cart
The Small Microfiber Swab has a polypropylene handle with a soft, non-abrasive, paddle-shaped tip. This absorbent swab tip is resistant to normal cleaning solvents, making it an ideal swab for removing contaminants and excess materials in fine assembly environments.
  • The PurSwab™ 3 in. small microfiber swab has a polypropylene handle with a soft, non-abrasive, paddle-shaped tip.
  • The absorbent swab tip is resistant to normal cleaning solvents, making it an ideal swab for removing contaminants and excess materials in fine assembly environments.
    • Low in particulates
    • Lint and residue free
    • Soft, absorbent, non-abrasive tip
    • Can be used with IPA, acetone, or other solvents
    • Low in NVRs
    • Used for applying adhesives or other fluids
    • Effective for removing residues and particulates
    • Ideal for fiber optics component manufacturing
    • All-around cleaning swab for controlled environments
    • Made in USA
Specifications
Length (English) 3 in.
Product Type Cleanroom Swab
Autoclavable Not Autoclavable
Sterility Non-sterile
Adhesive Type Ultrasonically Sealed
Handle Material Polypropylene
Tip Style Flex Paddle
Tip Material Microfiber