Learn More
Description
- Baked gray enamel metal cabinet
- Adjustable ozone output control provides from 3 to 300 mg/hr.
- Used in small to medium size laboratories: Up to 10000 cubic feet
- Eliminates various odors
- 5-year warranty included.
- The dimensions measure (10.5 L x 6.5 W x 8 H) in.
- Available with the FS017 Disposable Sporax Antimicrobial Replacement Filter as an optionalaccessory.
Specifications
Specifications
| For Use With (Equipment) | Enclosure |
| Product Type | Air Purifier |
| Voltage | 120 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
