Learn More
Memmert Memmert UN 75 PA, 115 Volt - Paraffin Oven. Maximum temperature range 20C-80C
SDP
Catalog No. 5000451129
Encompass_Preferred
Memmert UN 75 PA, 115 Volt - Paraffin Oven. Maximum temperature range 20C-80C
Catalog No. 50-004-51129 Supplier Memmert Supplier No. UN 75PA 115V
Memmert UN 75 PA, 115 Volt - Paraffin Oven. Maximum temperature range 20C-80C