Learn More
Chemglass Life Sciences Chemglass Small Polypropylene Process Reactor Funnel, 4.3 L, 260 mm Top, 80 mm Flange
SDP
Supplier:  Chemglass Life Sciences CG1767R810
Encompass_Preferred
Small polypropylene funnel designed for process reactor and powder transfer applications. It features a wide 260 mm top opening, an 80 mm reaction flange, and a 4.3 L capacity suitable for moderate batch operations.
  • Polypropylene construction for chemical resistance and durability.
  • 4.3 L capacity for handling moderate volumes.
  • 260 mm top diameter for wide-mouth filling.
  • 80 mm reaction flange for secure reactor connection.
  • Handle at rim for easier positioning and pouring.
Catalog No. NC2882263