Learn More
Electronic Imaging Materials Inc 626 XyResist chemical-resistant polyester slide labels, 1.00 inch x 0.75 inch, perforated, 6250 per roll, 3 inch core
SDP
Catalog No. NC9402042
Encompass_Preferred
1.00 inch x 0.75 inch, 626 XyResist chemical-resistant labels. Designed for harsh laboratory environments. Gloss topcoat is perfect for high resolution printing and provides extreme solvent, chemical, smudge, moisture and abrasion resistance when printed with T84 resin ribbon. It performs exceptionally well in chemical immersion tests—the adhesive bond easily handles Isopropyl Alcohol, Acetone, Bleach, and Xylene.
Catalog No. NC9402042 Supplier Electronic Imaging Materials Inc Supplier No. 626100075PR
1.00 inch x 0.75 inch, 626 XyResist chemical-resistant labels. Designed for harsh laboratory environments. Gloss topcoat is perfect for high resolution printing and provides extreme solvent, chemical, smudge, moisture and abrasion resistance when printed with T84 resin ribbon. It performs exceptionally well in chemical immersion tests—the adhesive bond easily handles Isopropyl Alcohol, Acetone, Bleach, and Xylene.