Learn More
Bellco Glass, Inc. Small Cell Scraper
SDP
Catalog No. 501946079
Encompass_Preferred
Stainless steel blade holder mounted on plastic covered stainless steel handle and is fully autoclavable. 5/8 X 1/4" BLADE X 7 3/4" HANDLE.. MADE IN THE USA Size: small
Catalog No. 50-194-6079 Supplier Bellco Glass, Inc. Supplier No. 773122005
Stainless steel blade holder mounted on plastic covered stainless steel handle and is fully autoclavable. 5/8 X 1/4" BLADE X 7 3/4" HANDLE.. MADE IN THE USA Size: small