Learn More
New England Biolabs, Inc. NEBNext Multiplex Small RNA Library Prep Set for Illumina (Set 1) – 24 reactions
SDP
Catalog No. NC0477293
Encompass
The novel NEBNext Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries. The NEBNext Small RNA Library Prep Set for Illumina (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.
Catalog No. NC0477293 Supplier New England Biolabs, Inc. Supplier No. E7300S
The novel NEBNext Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries. The NEBNext Small RNA Library Prep Set for Illumina (Set 1) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.