Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Small Serological Pipette Holder - 3 Pocket
SDP

Catalog No. 01259164
Change view
Click to view available options
Dimensions (L x W x H):
11.5 x 3.3 x 12 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-259-164 11.5 x 3.3 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
N/A
Catalog No. 01-259-164 Supplier Poltex Supplier No. CTUSRPIP3
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Small serological pipette holder with 3 pocket low profile design for tight spaces. Raised for laminar flow underneath with durable and cleanable PETG construction.

  • Small serological pipette holder
  • 3-pocket design
  • Low-profile for tight spaces or minimal pipette storage needs
  • A raised base allows laminar flow underneath
  • Durable and cleanable PETG construction
  • Consumables not included

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Serological Pipets, Pipets, Glass and Plastic Pipets, Serological Pipets (4 Types)
Material PETG
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Application) Serological Pipette Holder
For Use With (Equipment) Pipettes
Quantity 1 Ea.
Dimensions (L x W x H) 11.5 x 3.3 x 12 in.
Weight (English) 3.5 lb.
Product Type Serological Pipet Holder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.