Learn More
Newcomer Supply Slide Warmer with Cover, 10" x 7" Surface with digital controller
SDP
Catalog No. NC1613457
Encompass
Slide Warmer with Cover is ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Features: thermal heater ensures even heat transfer, anodized black surface provides contrast, LED temperature display, clear plastic lid allows you to clearly see your specimens and add humidity during your heating, thermostat setting range from room temperature to 75°C (+/- 2°C). Approximate capacity is 23 slides, weight is 10 lbs and power requirements 100W. CE certified and cord is CSA & UL listed.
Catalog No. NC1613457 Supplier Newcomer Supply Supplier No. 6262
Slide Warmer with Cover is ideal for use in the fields of cytology, histology, pathology, and biology for paraffin tissue section mounting. Features: thermal heater ensures even heat transfer, anodized black surface provides contrast, LED temperature display, clear plastic lid allows you to clearly see your specimens and add humidity during your heating, thermostat setting range from room temperature to 75°C (+/- 2°C). Approximate capacity is 23 slides, weight is 10 lbs and power requirements 100W. CE certified and cord is CSA & UL listed.