Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Small Steel Drum Funnel For Use With 5-Gal Steel Pail

Catalog No. 19822448 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
5 Gallon Steel Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-822-448 5 Gallon Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-822-448 Supplier Justrite™ Supplier No. 08201

Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Small Steel Drum Funnel For Use With 5-Gal Steel Pail is used to prevent spills when transferring flammable solvents, thinners, oils, or paint wastes.

Small Steel Funnel with Self-closing Cover and 1 in. Flame Arrester. For use with 5 gal. steel pails with 2 in. NPT.

  • Meets OSHA 29 CFR1910.1030 for the segregation of infectious waste
  • Perfect for medical facilities and research laboratories

Specifications

Description Steel Drum Funnel with Self-closing Cover
Color Yellow
For Use With (Equipment) 5 Gallon Steel Pail
Material Galvanized Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.