Learn More
Med Vet International Small Stifle Distractor w/Spinlock, 5in, J0240WAS, Each
Catalog No. 5000585852
Encompass_Preferred
The working tips on our spinlock are the same as the ratchet version. The spinlock offers more secure distraction as it is less likely to slip. The spinlock also offers an infinite range of position versus the ratchet.
Catalog No. 50-005-85852 Supplier Med Vet International Supplier No. JOR240WAS
The working tips on our spinlock are the same as the ratchet version. The spinlock offers more secure distraction as it is less likely to slip. The spinlock also offers an infinite range of position versus the ratchet.