Learn More
World Precision Instrument Small Syringe Multi Rack for SPLG pumps
SDP
Catalog No. 502539335
Encompass
A multi-rack system on a syringe pump gives you options. A syringe pump is a device used to precisely control the infusion or withdrawal of fluids. The multi-rack system allows the syringe pump to hold and control multiple syringes simultaneously. This is particularly useful when there is a need for infusing multiple fluids or drugs into a system at different rates or volumes. Each syringe in the rack can be independently controlled, enabling more complex experimental setups. This can save time and streamline experimental processes. You can ensure precise and synchronized delivery when you are doing continuous, simultaneous infusion of multiple solutions. The ability to use different syringe sizes and types in the same pump setup provides flexibility in experimental design. Researchers can choose syringes of varying capacities based on the specific requirements of each infusion.
Catalog No. 50-253-9335 Supplier World Precision Instrument Supplier No. 504576
A multi-rack system on a syringe pump gives you options. A syringe pump is a device used to precisely control the infusion or withdrawal of fluids. The multi-rack system allows the syringe pump to hold and control multiple syringes simultaneously. This is particularly useful when there is a need for infusing multiple fluids or drugs into a system at different rates or volumes. Each syringe in the rack can be independently controlled, enabling more complex experimental setups. This can save time and streamline experimental processes. You can ensure precise and synchronized delivery when you are doing continuous, simultaneous infusion of multiple solutions. The ability to use different syringe sizes and types in the same pump setup provides flexibility in experimental design. Researchers can choose syringes of varying capacities based on the specific requirements of each infusion.