Learn More
Ampco Small Valve Wheel Hook
SDP
Catalog No. 19175144 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
17 -3/4 in.
Catalog No. Length (English)
19-175-144 17 -3/4 in.
1 options
Catalog No. 19-175-144 Supplier Ampco Supplier No. WV36
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant valve wheel hook. Made in the USA.
  • F-type valve wheel hook
  • Aluminum bronze material, beryllium free.
  • Includes lifetime warranty.
  • Please see image for more information regarding measurements.
  • 17 and 3/4 in. length.
  • a (jaw opening size): 1 and 3/4 in.
  • b (throat depth): 3 in.
Specifications
Product Type Valve Wheel Lock
Length (English) 17 -3/4 in.
Quantity 1 Ea.