Learn More
Andwin Scientific SMALL WEIGHING BOATS 500/PK
SDP
Catalog No. NC9653914
Encompass_Preferred
The plastic weighing boats are manufactured to aid in weighing liquid or solid samples. This product is designed with recessed corners to allow controlled pouring. The boats have a square shape and a contour bottom.
Catalog No. NC9653914 Supplier Andwin Scientific Supplier No. 70041
The plastic weighing boats are manufactured to aid in weighing liquid or solid samples. This product is designed with recessed corners to allow controlled pouring. The boats have a square shape and a contour bottom.