Learn More
World Precision Instrument Smallest Fiber Optic Dipping Probe-DIP-UV-MINI-2
SDP
Catalog No. 502538870
Encompass
Mini DipTip™s  outer diameter is only 1.5 mm—the size of a 17-gauge needle. It will fit into all micro centrifuge tubes on the market and is a useful tool for measuring protein and DNA samples. The diminutive dipping probe can also be used for a dissolution system.
Catalog No. 50-253-8870 Supplier World Precision Instrument Supplier No. DIPUVMINI2
Mini DipTip™s  outer diameter is only 1.5 mm—the size of a 17-gauge needle. It will fit into all micro centrifuge tubes on the market and is a useful tool for measuring protein and DNA samples. The diminutive dipping probe can also be used for a dissolution system.