Learn More
Creative Biomart Recombinant Influenza B virus (Malaysia) Antigen
Catalog No. 502085131
Encompass_Preferred
Recombinant Full-Length B/Malaysia/2506/2004 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells. The MW is approximately 72,000 Dalton.
Catalog No. 50-208-5131 Supplier Creative Biomart Supplier No. VAC0291
Recombinant Full-Length B/Malaysia/2506/2004 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells. The MW is approximately 72,000 Dalton.