Learn More
CUBE BIOTECH SMALP 140; 1 g
Catalog No. NC2329198
Encompass
The synthethic polymer SMALP 140, among our other synthetic nanodisc products has been developed for for the solubilization & subsequent purification of membrane proteins from all kinds of cell membranes. Lyophilized Powder, to be solved with water / M.W. 5,000 Da
Catalog No. NC2329198 Supplier CUBE BIOTECH Supplier No. 18221
The synthethic polymer SMALP 140, among our other synthetic nanodisc products has been developed for for the solubilization & subsequent purification of membrane proteins from all kinds of cell membranes. Lyophilized Powder, to be solved with water / M.W. 5,000 Da