Learn More
CUBE BIOTECH SMALP 200; 1 g
Supplier:  CUBE BIOTECH 18211
Encompass
The SMALP, among all synthetic nanodisc products has been developed for for the solubilization of membrane proteins from all kinds of cell membranes and subsequent isolation & purification of said membrane proteins. After dissolving SMALP 200 + buffer powder in dH2O, the product is ready-to-use. SMALP 200 has a yellowish color in solution. / Lyophilized Powder, to be solved with water / M.W. 6,500 Da
Catalog No. NC2329208