Learn More
CUBE BIOTECH SMALP BZ Screening Kit; 1 kit
Catalog No. NC2682773
Encompass
3 tubes each containing 50 mg of four different nanodisc copolymers. Namely SMALP BZ25, BZ30, BZ35. BZ40. For the stabilization of membrane proteins outside the cell
Catalog No. NC2682773 Supplier CUBE BIOTECH Supplier No. 16690
3 tubes each containing 50 mg of four different nanodisc copolymers. Namely SMALP BZ25, BZ30, BZ35. BZ40. For the stabilization of membrane proteins outside the cell