Learn More
CUBE BIOTECH SMALP BZ25; 1 g
Catalog No. NC2684578
Encompass
1g of nanodisc copolymer SMALP BZ25 for the stabilization of membrane proteins outside the cell
Catalog No. NC2684578 Supplier CUBE BIOTECH Supplier No. 18612
1g of nanodisc copolymer SMALP BZ25 for the stabilization of membrane proteins outside the cell