Learn More
CUBE BIOTECH SMALP BZ30; 1 g
Catalog No. NC2684579
Encompass
1g of nanodisc copolymer SMALP BZ30 for the stabilization of membrane proteins outside the cell
Catalog No. NC2684579 Supplier CUBE BIOTECH Supplier No. 18622
1g of nanodisc copolymer SMALP BZ30 for the stabilization of membrane proteins outside the cell