Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Smap-2 (DT-1154) | 1809068-70-9 | 99.5% | 10 MG
SDP

Catalog No. 5000234495
Encompass_Preferred

SMAP-2 (DT-1154) is an orally active small-molecule activator of protein phosphatase 2A (PP2A) that exhibits anti-cancer activity in cell and animal models, reducing viability of pancreatic ductal adenocarcinoma cell lines and demonstrating in vivo efficacy.

  • Orally active PP2A activator with demonstrated anti-cancer activity.
  • High purity (99.5%).
  • Molecular weight 532.57 g/mol; formula C27H27F3N2O4S.
  • High solubility in DMSO (300 mg/mL) and formulatable for in vivo dosing using recommended protocols.
  • Stable as a powder when stored at -20°C and 4°C for indicated durations.
  • For research use only; not for human use.

Catalog No. 50-002-34495 Supplier Medchemexpress LLC Supplier No. HY12027210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMAP-2 (DT-1154) is an orally active small-molecule activator of protein phosphatase 2A (PP2A) that exhibits anti-cancer activity in cell and animal models, reducing viability of pancreatic ductal adenocarcinoma cell lines and demonstrating in vivo efficacy.

  • Orally active PP2A activator with demonstrated anti-cancer activity.
  • High purity (99.5%).
  • Molecular weight 532.57 g/mol; formula C27H27F3N2O4S.
  • High solubility in DMSO (300 mg/mL) and formulatable for in vivo dosing using recommended protocols.
  • Stable as a powder when stored at -20°C and 4°C for indicated durations.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.