Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Smap-2 (DT-1154) | 1809068-70-9 | 99.5% | 5 MG
SDP

Catalog No. 5000234567
Encompass_Preferred

SMAP-2 (DT-1154) is a small-molecule, orally active activator of protein phosphatase 2A (PP2A) with demonstrated anti-cancer activity in preclinical studies. It is supplied as a white to off-white solid and includes manufacturer-reported solubility and storage guidance.

  • Orally bioavailable PP2A activator with anticancer activity.
  • High purity: 99.53% (manufacturer reported).
  • Molecular weight 532.57; formula C27H27F3N2O4S.
  • Soluble in DMSO (300 mg/mL); in vivo formulation guidance provided.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years).

Catalog No. 50-002-34567 Supplier Medchemexpress LLC Supplier No. HY1202725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SMAP-2 (DT-1154) is a small-molecule, orally active activator of protein phosphatase 2A (PP2A) with demonstrated anti-cancer activity in preclinical studies. It is supplied as a white to off-white solid and includes manufacturer-reported solubility and storage guidance.

  • Orally bioavailable PP2A activator with anticancer activity.
  • High purity: 99.53% (manufacturer reported).
  • Molecular weight 532.57; formula C27H27F3N2O4S.
  • Soluble in DMSO (300 mg/mL); in vivo formulation guidance provided.
  • Powder storage: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.