Learn More
Cayman Chemical SMAP 29 (ovine) (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502916207
Encompass_Preferred
An antimicrobial peptide; bactericidal to P. aeruginosa strain PAO1 under low- and high-salt conditions in an energy-dependent luminescence assay (EC50s = 0.05 and 0.06 µM, respectively)
Catalog No. 50-291-6207 Supplier Cayman Chemical Supplier No. 3746210 MG
An antimicrobial peptide; bactericidal to P. aeruginosa strain PAO1 under low- and high-salt conditions in an energy-dependent luminescence assay (EC50s = 0.05 and 0.06 µM, respectively)