Learn More
Aviva Systems Biology Corp SMAP1 Antibody - middle region (ARP56282_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56282_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMAP1. Alternate names: FLJ13159, FLJ42245, SMAP-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61795