Learn More
Aviva Systems Biology Corp SMARCA1 Antibody - N-terminal region (ARP34512_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34512_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCA1. Alternate names: DKFZp686D1623, FLJ41547, ISWI, NURF140, SNF2L, SNF2L1, SNF2LB, SWI, SWI2, SNF2LTApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55431