Learn More
ABclonal Technology SMARCA2 / BRM Rabbit mAb
SDP
Supplier:  ABclonal Technology A23291100UL
Encompass_Preferred
The protein encoded by this gene is a member of the SWI SNF family of proteins and is highly similar to the brahma protein of Drosophila Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF SWI which is required for transcriptional activation of genes normally repressed by chromatin Alternatively spliced transcript variants encoding different isoforms have been found for this gene which contains a trinucleotide repeat (CAG) length polymorphism
Catalog No. 50-000-06465