Learn More
Aviva Systems Biology Corp SMARCA3 Antibody - C-terminal region (ARP38218_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38218_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SMARCA3. Alternate names: ZBU1, HLTF1, RNF80, HIP116, SNF2L3, HIP116A, SMARCA3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-56569