Learn More
Aviva Systems Biology Corp SMARCA5 Antibody - N-terminal region (ARP38368_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38368_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCA5. Alternate names: ISWI, SNF2H, WCRF135, hISWI, hSNF2HApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-014-56625