Learn More
Aviva Systems Biology Corp SMARCA5 Antibody - N-terminal region (ARP30031_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30031_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCA5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCA5. Alternate names: ISWI, SNF2H, WCRF135, hISWI, hSNF2HApplicationCHIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54105