Learn More
Aviva Systems Biology Corp SMARCAD1 Antibody - N-terminal region (ARP34592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCAD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCAD1. Alternate names: DKFZp762K2015, ETL1, KIAA1122, HEL1, ADERMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-55468