Learn More
Aviva Systems Biology Corp SMARCB1 Antibody - middle region (ARP34172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SMARCB1. Alternate names: RDT, CSS3, INI1, SNF5, Snr1, BAF47, MRD15, RTPS1, Sfh1p, hSNFS, SNF5L1, SWNTS1, PPP1R144ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55279