Learn More
Aviva Systems Biology Corp SMARCB1 Antibody - N-terminal region (ARP34171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCB1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human SMARCB1. Alternate names: RDT, INI1, SNF5, Snr1, BAF47, RTPS1, Sfh1p, hSNFS, SNF5L1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-55278