Learn More
Aviva Systems Biology Corp SMARCC1 Antibody - C-terminal region (ARP33307_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33307_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SMARCC1. Alternate names: Rsc8, SRG3, SWI3, BAF155, CRACC1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54954