Learn More
Aviva Systems Biology Corp SMARCD2 Antibody - C-terminal region (ARP38223_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SMARCD2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SMARCD2. Alternate names: BAF60B, CRACD2, PRO2451, Rsc6pApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56570