Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ SMART 3™ Colorimeter Water Quality Analyzer
GSA_VASDPSCI_ED

Catalog No. S66155A Shop All LaMotte Company Products
Change view
Click to view available options
Wavelength Range:
428, 520, 568, 635 nm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength Range
S66155A 428, 520, 568, 635 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S66155A Supplier LaMotte™ Supplier No. 1910
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Six sample tubes, USB cable, USB wall plug, instruction manual and a lithium rechargeable battery. Reagent systems for use with colorimeter are sold separately.

Multitest colorimeter measures numerous water quality parameters with laboratory accuracy and convenience of a portable, hand-held system.

Captures up to 500 date- and time-stamped data points with the datalogger, then download via USB connection.

  • Compact unit with large, backlit display
  • More than 80 pre-programmed tests
  • Automatic wavelength selection
  • Menu-driven operation
  • Accepts COD vials and cuvets
  • Waterproof to IP67
  • Seven languages — user selectable
  • USB port and cable for downloading results and charging
  • Multiple LED optical system
  • Option of entering up to 25 additional calibrations for analysis with custom reagent systems

Order Info

Accessories, such as SMARTLink 3 Software (S66155B), replacement USB cable (S66155C) and COD/UDV adapter (S66155D), are available separately. LaMotte reagent systems for use with SMART 3 Colorimeter sold separately.

Certifications

CE marked

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accuracy ±0.005 AU at 1 AU
Calibration Yes
Certifications/Compliance CE, EPA
Connections Mini B USB
Battery Type Rechargeable Lithium-Ion
Detector Type Photodiode
Display Backlit LCD
Dimensions (L x W x H) 3.5 x 7.5 x 2.5 in. (8.84 x 19.05 x 6.35 cm)
IP Rating IP67
Languages English, French, Spanish, Italian, Portuguese, Chinese, Japanese
Light Source LED
Spectral Bandwidth 10 nm
Type Digital Colorimeter
Voltage 3.7 V
Wavelength Range 428, 520, 568, 635 nm
Wavelength Selection Automatic
For Use With (Application) Industrial water boiling and cooling, water and wastewater
Height (English) 2.5 in.
Height (Metric) 6.35 cm
Includes Six sample tubes, USB cable, USB wall plug, instruction manual and a Lithium rechargeable battery
Length (English) 3.5 in.
Length (Metric) 8.84 cm
Resolution 1% FS
Warranty 2 Years
Weight (English) 13 oz.
Weight (Metric) 362 g
Width (English) 7.5 in.
Width (Metric) 19.05 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.