Learn More
MSC First Aid Only Smart Compliance First Aid Kit, 165 Pieces, Wall Mount, 25 Person Capacity, 1 ea
Catalog No. 5000685244
Encompass
This 165-piece wall-mounted first aid kit provides essential supplies to cleanse and treat minor injuries in work environments of up to 25 people. Designed for convenience and safety, the kit features a grid-based storage system that allows individual boxes to snap in easily for organized access.
  • Contains 165 pieces to support up to 25 people
  • Includes individual packets for easy use and to help prevent cross-contamination
  • Effective for cleansing and preventing infection in minor wounds, burns, and scrapes
  • Includes bandages suitable for minor cuts, abrasions, and puncture wounds
  • Equipped with benzalkonium chloride for wound care
  • Mounts securely onto flat vertical surfaces using strong adhesive tape
  • Features re-orderable boxes that snap directly into the wall-mounted grid
Catalog No. 50-006-85244 Supplier MSC Supplier No. 16341752
This 165-piece wall-mounted first aid kit provides essential supplies to cleanse and treat minor injuries in work environments of up to 25 people. Designed for convenience and safety, the kit features a grid-based storage system that allows individual boxes to snap in easily for organized access.
  • Contains 165 pieces to support up to 25 people
  • Includes individual packets for easy use and to help prevent cross-contamination
  • Effective for cleansing and preventing infection in minor wounds, burns, and scrapes
  • Includes bandages suitable for minor cuts, abrasions, and puncture wounds
  • Equipped with benzalkonium chloride for wound care
  • Mounts securely onto flat vertical surfaces using strong adhesive tape
  • Features re-orderable boxes that snap directly into the wall-mounted grid