Learn More
Martor Secunorm Smartcut W/ Stainless Steel Blade
Catalog No. NC9023056
Encompass_Preferred
The Smartcut is a disposable, semi automatic retractable spring loaded knife for cutting general industrial materials.-Semi automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of aluminum-Safer alternative to fixed blade utility knives for general cutting tasks
Catalog No. NC9023056 Supplier Martor Supplier No. 110100.08
The Smartcut is a disposable, semi automatic retractable spring loaded knife for cutting general industrial materials.-Semi automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of aluminum-Safer alternative to fixed blade utility knives for general cutting tasks