Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SMART Digest™ Chymotrypsin Kits

Catalog No. 03252211 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Bulk Resin option
Bulk Resin option with Filter/Collection plate
Bulk Resin option with SOLAμ/collection plate
Low pH, Magnetic Beads, Bulk option for MAM workflows
Magnetic Bulk Resin option
Magnetic Bulk Resin option with Filter/Collection plate
Magnetic Bulk Resin option with SOLAμ/collection plate
SMART Digest Chymotrypsin Kit with SOLAμ/collection plate
with Collection plate
with Filter/Collection plate
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
03-252-211 Magnetic Bulk Resin option with SOLAμ/collection plate
03-251-707 with Collection plate
03-251-901 Low pH, Magnetic Beads, Bulk option for MAM workflows
03-252-207 Bulk Resin option
03-252-208 Magnetic Bulk Resin option
03-251-708 with Filter/Collection plate
03-252-209 Bulk Resin option with Filter/Collection plate
13-622-453 Magnetic Bulk Resin option with Filter/Collection plate
03-251-709 SMART Digest Chymotrypsin Kit with SOLAμ/collection plate
03-252-210 Bulk Resin option with SOLAμ/collection plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 03-252-211 Supplier Thermo Scientific™ Supplier No. 60109106MB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obtain fast, simple, and highly reproducible chymotrypsin protein digestion for peptide characterization and quantitation in biopharmaceutical research.

Obtain high-quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Chymotrypsin Kits. A significant advance in sample preparation for biopharmaceutical protein research, these kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. SMART Digest Kits are a significant improvement over in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.

Chymotrypsin Protein Digestion

Characterize and quantitate proteins faster, easier, and quicker than ever before using SMART Digest kits. This version of the kit uses chymotrypsin, which cleaves peptide bonds formed by aromatic residues such as tyrosine, phenylalanine, and tryptophan.

Easy and Simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time

Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digestion can take up to 24 hours.

Cleaner Samples

Reduction in the number of chemicals used and the immobilized chymotrypsin design helps mitagate against chemically induced post translational modifications (PTMs) and autolysis. The results are easier to interpret.

Couple with SOLAμ SPE plates for desalting

When used with the SMART Digest for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.

TRUSTED_SUSTAINABILITY

Specifications

Description Magnetic Bulk Resin option with SOLAμ/collection plate
Capture None
Cleavage Peptide bonds of aromatic residues (e.g., tyrosine, phenylalanine, tryptophan)
Enzyme Chymostrypsin
Format SMART Digest Resin, Bulk Magnetic
For Use With (Application) Peptide Quantitation, Peptide Mapping
Product Line SMART Digest
Type Protein Digestion
Includes Bulk Magnetic SMART Digest Resin, SMART Digest Buffer, Collection Plate, SOLAμ SPE Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.