Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SMART Digest™ OligoSelect™ Kit (Magnetic WAX Bead Format)

Catalog No. 03050600 Shop All Thermo Scientific Products
Change view
Click to view available options
Enzyme:
Proteinase K
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Enzyme
03-050-600 Proteinase K
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03-050-600 Supplier Thermo Scientific™ Supplier No. 60190101
Add to Cart
Add to Cart

Thermo Scientific™ SMART Digest™ OligoSelect Kits enable efficient disruption of protein-nucleotide binding and extraction of oligonucleotide-based drugs and metabolites from biofluids and tissues for accurate quantitation by LC-MS. This kit includes all necessary enzymes, buffers, and solid phase extraction (SPE) materials.

The SMART Digest™ OligoSelect Kit provides a streamlined solution for the disruption of protein-nucleotide binding and extraction of oligonucleotide-based drugs and metabolites from biofluids and tissues, ensuring precise quantitation by LC-MS. This kit includes all essential enzymes, buffers, and extraction materials, offering a comprehensive and reliable choice for your laboratory needs.

  • Efficiently disrupts protein-nucleotide binding and extracts oligonucleotide-based drugs and metabolites from biofluids
  • Ensures precise quantitation by LC-MS
  • Includes all necessary enzymes for effective protein digestion
  • Comes with required buffers for optimal enzyme activity
  • Features magnetic WAX beads for high-throughput sample processing
  • Comprehensive kit designed to streamline your workflow
TRUSTED_SUSTAINABILITY

Specifications

Bed Weight 5 mg
Capture None
Cleavage Proteinase K preferentially cleaves peptide bonds at the carboxylic side of aliphatic and aromatic amino acids
Enzyme Proteinase K
Format Magnetic
For Use With (Application) Efficient disruption of protein-nucleotide binding followed by extraction of oligonucleotide-based drugs and metabolites from biofluids and tissues prior to LC-MS quantitation.
For Use With (Equipment) Universal
Particle Size 30 to 70 μm
Product Line SMART Digest
Sorbent Mixed Mode Weak Anion Exchanger, Polymeric
Type Oligo Isolation
Volume (Metric) 1 mL
Volume (Metric) Well 1 mL
Includes SMART Digest Soluble Proteinase K (0.5 mL 20 mg/mL), Proteinase K Digest Buffer (25 mL), Magnetic WAX beads 96 well (5 mg/well), WAX Elution Buffer (25 mL)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.