Learn More
STEMCELL Technologies SmartDish™ - 50 Pack, Size: 50 Units
SDP
Catalog No. NC1749673
Encompass
Meniscus-free cultureware for more accurate counting of hematopoietic colonies in CFU assays
This item is not returnable. View return policy
Catalog No. NC1749673 Supplier STEMCELL Technologies Supplier No. 27371
Meniscus-free cultureware for more accurate counting of hematopoietic colonies in CFU assays