Learn More
Thomas Scientific SmartDoc Band Pass Filter, 535nm, for imaging Green Stains on UV transilluminator
SDP
Catalog No. 501459548
Encompass
Ideal for common safe stains (SmartGlow™, SYBR Green, GelGreen™, etc.) Visible blue light will not damage DNA Quality gel imaging with any smart phone Gel access for DNA band cutting Imaging enclosure can also be used on existing transilluniators Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV, which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with safe green dyes (SYBR Green, SmartGlow™, GelStar™, GelGreen™, etc). The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands
Catalog No. 50-145-9548 Supplier Thomas Scientific Supplier No. 1177X53
Ideal for common safe stains (SmartGlow™, SYBR Green, GelGreen™, etc.) Visible blue light will not damage DNA Quality gel imaging with any smart phone Gel access for DNA band cutting Imaging enclosure can also be used on existing transilluniators Introducing the SmartDoc 2.0. This complete system has been developed for safely viewing nucleic acids in agarose gels and also for capturing quality digital images with a smart phone camera. Blue light is used instead of UV, which is safer for lab personnel and will not damage DNA samples. The illumination base emits an intense blue light with a peak output of 460nm for optimum excitation of nucleic acids stained with safe green dyes (SYBR Green, SmartGlow™, GelStar™, GelGreen™, etc). The orange cover fits on top of the illumination base and acts as a filter to absorb blue light, allowing visualization of the fluorescing samples. The cover can also be set at an angle for gel access and cutting out DNA bands