Learn More
Takara Bio SMARTer® RACE 5’/3’ Kit, 10 Rxns
SDP
Supplier:  Takara Bio 634858
Encompass
The SMARTer RACE 5'/3' Kit allows the synthesis of first-strand cDNA from poly A+ or total RNA via SMART (Switching Mechanism At 5' End of RNA Template) technology, and facilitates the performance of 5'- and 3'-RACE (Rapid Amplification of cDNA Ends) PCR with the kit's Universal Primer Mix. Our carefully designed, specially-modified SMARTer Oligo preferentially captures the 5' ends of the cDNA during cDNA synthesis. Using this SMARTer Oligo, our procedure enriches cDNA pools for 5' sequences, thus increasing the likelihood you will amplify the entire sequence of your gene. The SMARTer RACE 5'/3' Kit has been improved to accommodate larger RNA input volumes and perform better on challenging targets than the original SMARTer RACE cDNA Amplification Kit. The In-Fusion HD Cloning Kit, NucleoSpin Gel and PCR Clean-Up Kit, and Stellar Competent Cells are included for your convenience in cloning RACE products. 10 Rxns
Catalog No. NC0745266