Learn More
MilliporeSigma™ Smart Flare™ SOX2; Human, Cyanine3 RNA Detection Probe
Catalog No. SF914 Shop All MilliporeSigma Products
Change view
Click to view available options
SOX2, Human, Cyanine3 RNA Detection Probe
Catalog No.
SF914 SOX2, Human, Cyanine3 RNA Detection Probe
1 options
Catalog No. SF914 Supplier MilliporeSigma™ Supplier No. SF914
Used as biomarker to detect sequence specfic RNA.
  • Fluorescent reagent, enables sequence specific mRNA
  • MicroRNA expression detection at single cell resolution
  • Allows researchers to identify cells expressing biomarkers of interest
  • Enrich sort cells based on individual expression levels through FACS
  • Compatible with downstream functional analysis