Learn More
Healthmark Industries Co SMART PACK LS 30/PK
SDP
Catalog No. NC2960900
Encompass_Preferred
  • Designed for daily operational checks of steam sterilizers
  • Calibrated for 11 minutes exposure time
  • Test temperature set at 121°C (250°F)
  • Complete color change to black indicates satisfactory results
  • Bowie-Dick type test ensures air removal efficiency
  • Lead and toxic heavy metals free composition
  • Single-use application for hygiene
  • Latex free for safety
  • Shelf life of 24 months
  • 30 test packs included
This item is not returnable. View return policy
Catalog No. NC2960900 Supplier Healthmark Industries Co Supplier No. 61301606325
  • Designed for daily operational checks of steam sterilizers
  • Calibrated for 11 minutes exposure time
  • Test temperature set at 121°C (250°F)
  • Complete color change to black indicates satisfactory results
  • Bowie-Dick type test ensures air removal efficiency
  • Lead and toxic heavy metals free composition
  • Single-use application for hygiene
  • Latex free for safety
  • Shelf life of 24 months
  • 30 test packs included