Learn More
Formulatrix SMART PUMP BOX V3
SDP
Catalog No. NC3512978
Encompass
  • Enables accurate dispensing across a broad range of liquid viscosities without consumables.
  • Facilitates rapid and efficient liquid transfers for scientific workflows.
  • Supports tip-free dispensing, reducing consumable costs and minimizing wastage.
  • Ensures consistent performance over millions of dispense cycles.
  • Handles diverse fluid types, including cells and beads, without clogging.
  • Automated dual-solution wash cycle for maintenance.
  • Compatible with SBS-footprint plates (6, 12, 24, 48, 96, 384, 1536-well).
  • Intuitive GUI with Excel import capability.
  • Easily integrated with robotic handlers.
Catalog No. NC3512978 Supplier Formulatrix Supplier No. 001455C
Add to Cart
Add to Cart
Request a Quote
  • Enables accurate dispensing across a broad range of liquid viscosities without consumables.
  • Facilitates rapid and efficient liquid transfers for scientific workflows.
  • Supports tip-free dispensing, reducing consumable costs and minimizing wastage.
  • Ensures consistent performance over millions of dispense cycles.
  • Handles diverse fluid types, including cells and beads, without clogging.
  • Automated dual-solution wash cycle for maintenance.
  • Compatible with SBS-footprint plates (6, 12, 24, 48, 96, 384, 1536-well).
  • Intuitive GUI with Excel import capability.
  • Easily integrated with robotic handlers.