Learn More
BEST SANITIZERS Smart-San Hand Sanitizer Foam is engineered to meet the tough requirements of repeated daily use while keeping hands soft and healthy. Effective against 30 pathogens. Available in 500ml pump bottle and 1.2L cartridge refill
Catalog No. NC3338785
Encompass_Preferred
Smart-San Hand Sanitizer Foam is engineered specifically to meet the tough requirements of repeated daily use while keeping hands soft and healthy. It is a powerful, yet gentle ethanol-based formula which has been tested and proven effective against 30 pathogens. Formulated with emollients to help enhance the skin's protective barrier while sanitizing. Available in 500ml pump bottle and 1.2L cartridge refill 12/CS
Catalog No. NC3338785 Supplier BEST SANITIZERS Supplier No. BHC1010
Smart-San Hand Sanitizer Foam is engineered specifically to meet the tough requirements of repeated daily use while keeping hands soft and healthy. It is a powerful, yet gentle ethanol-based formula which has been tested and proven effective against 30 pathogens. Formulated with emollients to help enhance the skin's protective barrier while sanitizing. Available in 500ml pump bottle and 1.2L cartridge refill 12/CS