Learn More
Research Products International Corp SmartSlicer Safety Razor Blade, 19 x 38 x 0.22mm Thick, 100 per Package
SDP
Catalog No. 501367935
Encompass
SmartSlicer provides a unique alternative to single edge razor blades. Made from ultra thin (0.22 mm) food grade PET plastic that performs like a conventional blade, however is much safer to use. Cleanly slice right on the light box without scratching the surface. Ideal for a variety of laboratory applications including slicing gels, soft tissue, plant matter and more. Adjust the size required by simply cutting with scissors. Made in the USA.
Catalog No. 50-136-7935 Supplier Research Products International Corp Supplier No. 246810
SmartSlicer provides a unique alternative to single edge razor blades. Made from ultra thin (0.22 mm) food grade PET plastic that performs like a conventional blade, however is much safer to use. Cleanly slice right on the light box without scratching the surface. Ideal for a variety of laboratory applications including slicing gels, soft tissue, plant matter and more. Adjust the size required by simply cutting with scissors. Made in the USA.