Learn More
LaMotte™ SMART Spectro™ Spectrophometer and SMART 3™ Colorimeter Reagent Systems
Shop All LaMotte Company Products
Description
- High-quality reagents manufactured from ACS raw materials for high reproducibility
- All systems are designed to overcome test interferences for fast and dependable results
Specifications
Specifications
| For Use With (Application) | Water testing and environmental studies |
| For Use With (Equipment) | SMART Spectro Spectrophometer and SMART 3 Colorimeter |
| No. of Tests | 50 |
| Range | 0 to 0.3ppm |
| Test Method | Eriochrome Cyanine R |
| Type | Aluminum |