Learn More
Trajan Scientific Smart SPME Arrow 1.10 Dk Gry 1Pk
Catalog No. 501920806
Encompass
Smart SPME Arrow 1.10mm: DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane) Phase thickness 120µm Phase length 20mm color code: dark grey pack of 1 pc
Catalog No. 501920806 Supplier Trajan Scientific Supplier No. SARR11DVB/CWR120/20P1
Smart SPME Arrow 1.10mm: DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane) Phase thickness 120µm Phase length 20mm color code: dark grey pack of 1 pc